1-Hexadecyl-3-methylimidazolium chloride
Catalog # ABCRAB289677.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
:
:343 g/mol
CAS Number:61546-01-8
Formula:C₂₀H₃₉ClN₂
MDL Number:MFCD03427608