
Aluminium sulphate hexadecahydrate
Supplier: Lach-ner S.R.O.
Certificates
About this item
CAS Number:16828-11-8
:
Formula:Al₂O₁₂S₃·16H₂O
MDL Number:MFCD00149137
:630.4 g/mol
Melting Pt:86 °C
Density:1.69 g/cm³ (20 °C)
EINECS:233-135-0
:GHS05
Storage Temperature:Ambient
Frequently Bought Together