Sodium hypophosphite monohydrate
Catalog # LACH30052AP0G1000
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1.000 g
- Seq:3047
Specifications
About this item
:
:105.99 g/mol
Melting Pt:90 °C
Density:0.8 g/cm³ (20 °C)
EINECS:231-669-9
CAS Number:10039-56-2
Formula:NaH₂PO₂·H₂O
MDL Number:MFCD06252379