Aluminium ammonium sulphate dodecahydrate
Catalog # LACH30162CP0G05001
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
Synonyms: Aluminium ammonium bis(sulphate) dodecahydrate, Ammonium alum, Ammonium aluminum sulphate dodecahydrate, Aluminum ammonium sulphatedodecahydrate
:
:453.33 g/mol
EINECS:232-055-3
:GHS07
CAS Number:7784-26-1
:MFCD00149958
Formula:AlNH₄(SO₄)₂·12H₂O
MDL Number:MFCD00051042
Boiling Pt:200
Melting Pt:93
Density:1.64 g/cm³ (20 °C)
Merck Index:13,00325