




Nickel(II) nitrate hexahydrate
Catalog # LACH30193
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:290.79 g/mol
Merck Index:12,06596
Boiling Pt:137 °C (1013 hPa)
Melting Pt:56.7 °C
Density:2.05 g/cm³ (20 °C)
EINECS:236-068-5
UN:2725
ADR:5.1,III
:GHS03
:GHS05
:GHS08
:GHS07
:GHS09
CAS Number:13478-00-7
Formula:Ni(NO₃)₂·6H₂O
MDL Number:MFCD00149805