D-(+)-Glucose monohydrate
Supplier: Lach-ner S.R.O.
Certificates
About this item
CAS Number:14431-43-7
:5996-10-1
Formula:C₆H₁₂O₆·H₂O
MDL Number:MFCD00149450
:198.17 g/mol
Melting Pt:83 °C
Storage Temperature:Ambient
EINECS:200-075-1
Frequently Bought Together