di-Potassium hydrogen phosphate
Catalog # LACH30060FP0G1000
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1.000 g
- Seq:3047
Specifications
About this item
:
:174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-834-5
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383