Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

L(+)-Tartaric acid

Irritant
Supplier:   Lach-ner S.R.O.
L(+)-Tartaric acid
Supplier:  Lach-ner S.R.O.
Certificates
L(+)-Tartaric acid
L(+)-Tartaric acid

2 Options Available Below

Product Details & Documents
Synonyms:  L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
  • CAS Number:87-69-4
  • Formula:C₄H₆O₆
  • MDL Number:MFCD00064207
  • Molecular Weight:150.09 g/mol
  • Merck Index:13,09158
  • Boiling Pt:399 °C (1013 hPa)
  • Melting Pt:168…170 °C
  • Density:1.759 g/cm³ (20 °C)
  • Flash Pt:150 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:201-766-0
  • REACH:01-2119537204-47
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 557,00 Kč
    557,00 Kč
    1 * 500 g
       557,00 Kč

  • Plastic bottle for solids
    1.000 g
    Log in for availability
    Log in for availability
    1 * 1.000 g
    1 * 1.000 g

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.