
Titanium(IV) chloride
Catalog # LACH30426
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:189.69 g/mol
Merck Index:13,09555
Boiling Pt:136 °C (1013 hPa)
Melting Pt:136.2 °C
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-441-9
UN:1838
ADR:8,II
:GHS05
CAS Number:7550-45-0
Formula:Cl₄Ti
MDL Number:MFCD00011267