
Dimethyl-trans-cyclohexane-1,4-dicarboxylate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3399-22-2
:
Formula:C₁₀H₁₆O₄
MDL Number:MFCD00063917
Melting Pt:67…70 °C
:200.23 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:67 to 70 °C
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together