
6-Chloroimidazo[1,2-a]pyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6188-25-6
:
Formula:C₇H₅ClN₂
MDL Number:MFCD05663807
:152.58 g/mol
Melting Pt:80…82 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together