
5-Bromoimidazo[1,2-a]pyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69214-09-1
:
Formula:C₇H₅BrN₂
MDL Number:MFCD08275698
:197.03 g/mol
Melting Pt:61…64 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together