Sample bags, Whirl-Pak™
: DELTALAB
These LDPE blend sterile sample bags are resistant and transparent that are designed for both solid and liquid samples and the plastic thickness depends on the model. These are unique bags with a one-piece seam, avoiding the possible risks of the loss of the corners bags. To assure a leakproof closure to maintain the sample integrity, close the closing wires three times.
- Ethylene oxide sterilised
- Suitable for food industry
- Non autoclavable
- Leakproof closure with several metallic rounded sticks
- The bags resist temperature up to 82 °C
Frequently Bought Together