Order Entry
Czech Republic
ContactUsLinkComponent
Carboys, wide neck, LDPE
Carboys, wide neck, LDPE
Catalog # SEMA3060
Supplier:  SEMADENI
undefined
Carboys, wide neck, LDPE
Catalog # SEMA3060
Supplier:  SEMADENI

Specifications

  • Capacity:
    10 l
  • W×D×H:
    300×140×400 mm
  • Neck-Ø:
    90 mm
  • Pk:
    1

Specifications

About this item

LDPE, translucent.

  • Good chemical resistance
  • Good temperature resistance

With silicone tubing, pinchcock and screw cap.