

Barium nitrate
Catalog # LACH30026AP0G0500
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Merck Index:13,00986
Melting Pt:∼592 °C
Density:∼3.23 g/cm³ (20 °C)
ADR:5.1,II
:261.34 g/mol
EINECS:233-020-5
UN:1446
:GHS03
:GHS07
Storage Temperature:Ambient
CAS Number:10022-31-8
Formula:Ba(NO₃)₂
MDL Number:MFCD00003442