Water
LACH20059AT0L0010
:
- Pack type:Plastic bottle
- Pk:10 L
- Seq:5214
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
:
:18.02 g/mol
:13,10098
:100 °C (1013 hPa)
:0 °C
:1 g/cm³ (4 °C)
:Ambient
:231-791-2
:7732-18-5
:H₂O
:MFCD00011332