Order Entry
Czech Republic
ContactUsLinkComponent
Sodium cyanide
Sodium cyanide
Catalog # LACH30122-CP0-G
Supplier:  Lach-ner S.R.O.
CAS Number:  
Sodium cyanide
Catalog # LACH30122-CP0-G
Supplier:  Lach-ner S.R.O.
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    50 kg
  • Seq:
    4418

Specifications

About this item

 :
Merck Index:13,08679
Boiling Pt:1496 °C (1013 hPa)
Melting Pt:563.9 °C
Density:1.595 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-599-4
UN:1689
ADR:6.1,I
 :49.01 g/mol
 :GHS06
 :GHS09
CAS Number:143-33-9
Formula:CNNa
MDL Number:MFCD00003523