Benzoic acid
Catalog # LACH10016CP0G0500
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:122.12 g/mol
Merck Index:13,01092
Boiling Pt:249 °C (1013 hPa)
Melting Pt:121…123 °C
Density:1.316 g/cm³ (20 °C)
Flash Pt:121 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-618-2
REACH:01-2119455536-33
:GHS07
CAS Number:65-85-0
Formula:C₇H₆O₂
MDL Number:MFCD00002398