Order Entry
Czech Republic
ContactUsLinkComponent
 
Health
Environment
Tetrachloroethylene
Catalog # LACH20055UT0M2500
Supplier:  Lach-ner S.R.O.
CAS Number:  
undefined
Health
Environment
Tetrachloroethylene
Catalog # LACH20055UT0M2500
Supplier:  Lach-ner S.R.O.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :165.83 g/mol
Merck Index:12,09332
Boiling Pt:121.1 °C (1013 hPa)
Melting Pt:–22 °C
Density:1.63 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-825-9
UN:1897
ADR:6.1,III
 :GHS08
 :GHS09
CAS Number:127-18-4
Formula:C₂Cl₄
MDL Number:MFCD00000834