4-Methyl-2-pentanone
Glass bottle, 1 L
: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK
- :108-10-1
- :(CH₃)₂CHCH₂COCH₃
- :MFCD00008938
- :100.16 g/mol
- :13,05227
- :117.4 °C (1013 hPa)
- :–84 °C
- :0.801 g/cm³ (20 °C)
- :16 °C
- :Ambient
- :203-550-1
- :1245
- :3,II
- Pack type:Glass bottle
- Pk:1 L