Magnesium chloride hexahydrate
Catalog # LACH30080FP0G1000
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1.000 g
- Seq:3047
Specifications
About this item
:
:203.3 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:117 °C
Density:1.57 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7791-18-6
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781