Sodium thiosulphate pentahydrate
Supplier: Lach-ner S.R.O.
Jump to:
About this item
- CAS Number:10102-17-7
- Formula:Na₂S₂O₃.5H₂O
- MDL Number:MFCD00149186
- Molecular Weight:248.19 g/mol
- Merck Index:12,08844
- Melting Pt:48 °C
- Density:1.67 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-867-5
2 Options Available Below
Product Details & Documents
- CAS Number:10102-17-7
- Formula:Na₂S₂O₃.5H₂O
- MDL Number:MFCD00149186
- Molecular Weight:248.19 g/mol
- Merck Index:12,08844
- Melting Pt:48 °C
- Density:1.67 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-867-5



