

Lithium hydroxide monohydrate
Supplier: Lach-ner S.R.O.
Certificates
About this item
CAS Number:1310-66-3
:
Formula:LiOH·1H₂O
MDL Number:MFCD00149772
:41.96 g/mol
Merck Index:13,05556
Boiling Pt:920 °C (1013 hPa)
Melting Pt:462 °C
Density:1.51 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-183-4
UN:2680
ADR:8,II
:GHS06
:GHS05
Frequently Bought Together