
Lithium chloride monohydrate
Catalog # LACH221430405050
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:60.41 g/mol
EINECS:231-212-3
:GHS07
Storage Temperature:Ambient
CAS Number:16712-20-2
:85144-11-2
Formula:ClLi·1H₂O
MDL Number:MFCD00149764