
Ammonium metavanadate
Catalog # LACH30124CP0G02501
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:116.98 g/mol
Merck Index:13,00572
Melting Pt:200 °C
Density:2.33 g/cm³ (20 °C)
EINECS:232-261-3
UN:2859
ADR:6.1,II
:GHS06
CAS Number:7803-55-6
Formula:NH₄VO₃
MDL Number:MFCD00011430