


Sodium sulphide hydrate, Technical Grade
Supplier: Lach-ner S.R.O.
Certificates
About this item
CAS Number:27610-45-3
:
Formula:Na₂S·xH₂O
MDL Number:MFCD00149183
:96.06 g/mol (anhy)
Merck Index:12,08829
Boiling Pt:∼920 °C (1013 hPa)
Melting Pt:50 °C
Density:∼1.43 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:215-211-5
UN:1849
ADR:8,II
REACH:01-2119513694-38
:GHS06
:GHS05
:GHS09
Frequently Bought Together