Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
di-Potassium disulphite
Potassium disulphite
Potassium disulphite
LACH30023CP0G1000
:
Lach-ner S.R.O.
:
Potassium disulphite
LACH30023CP0G1000
:
Lach-ner S.R.O.
:
Pack type:
Plastic bottle for solids
Pk:
1.000 g
Seq:
3047
:
:
222.33 g/mol
:
13,07729
:
190 °C
:
1.2 g/cm³ (20 °C)
:
240-795-3
:
GHS05
:
GHS07
:
16731-55-8
:
K₂O₅S₂
:
MFCD00167605