
2,2-Bis(4-hydroxycyclohexyl)propane (mixture of isomeres), Sigma-Aldrich®
Catalog # SIAL416517-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:240.39 g/mol
:M-Clarity™ quality level = MQ100
EINECS:201-244-2
:GHS07
CAS Number:80-04-6
Formula:C₁₅H₂₈O₂
MDL Number:MFCD00019334