Potassium hydroxide analytical reagent
Catalog # LACH10003AP0G00001
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
About this item
Synonyms: Potash caustic, Caustic potash
:
:56.11 g/mol
EINECS:215-181-3
UN:1813
REACH:01-2119487136-33
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553
Boiling Pt:1320
Melting Pt:360
Density:2.04 g/cm³ (20 °C)
ADR:8 II
Merck Index:12,07806