
(1-Ethoxycarbonylethyl)triphenylphosphonium bromide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:30018-16-7
:
Formula:C₂₃H₂₄BrO₂P
MDL Number:MFCD00031605
:443.33 g/mol
EINECS:250-002-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together