
4-Nitro-3-(trifluoromethyl)benzenesulphonyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:39234-83-8
:
Formula:C₇H₃ClF₃NO₄S
MDL Number:MFCD03094387
:289.62 g/mol
Melting Pt:63…65 °C
Storage Temperature:Ambient
ADR:8,II
:GHS05
Related Information
Frequently Bought Together