


4-Fluorphenylmagnesium bromide 2 mol/l in diethyl ether, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:352-13-6
:
Formula:C₆H₄BrFMg
MDL Number:MFCD00009667
Density:1.043 g/cm³
Flash Pt:–40 °C
UN:3399
ADR:4.3,I
:GHS07
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear to turbid yellow to brown Solution (May develop some turbidity or precipitate)
- Titration Grignard:1.9 to 2.2 M
Specification Test Results
Related Information
Frequently Bought Together