Order Entry
Czech Republic
ContactUsLinkComponent
5-(Trifluoromethoxy)salicylaldehyde ≥98%
Irritant
5-(Trifluoromethoxy)salicylaldehyde ≥98%
Catalog # L17136.03
CAS Number:  
Irritant
5-(Trifluoromethoxy)salicylaldehyde ≥98%
Catalog # L17136.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:93249-62-8
 :
Formula:C₈H₅F₃O₃
MDL Number:MFCD00075249
 :206.12 g/mol
Boiling Pt:82 °C (60 mmHg)
Melting Pt:31…33 °C
Flash Pt:>110 °C (230 °F)
 :GHS07
Storage Temperature:Ambient