4-Nitrophenol-[2,3,5,6-D4] 100 µg/ml in acetone, Dr Ehrenstorfer
EHERXA15590404AC
:
- Pack type:Glass bottle
- Pk:1,1 mL
- CAS No.:93951-79-2
- Description:4-Nitrophenol-[2,3,5,6-D4] 100 µg/ml in acetone
- Formula:C₆₂H₄HNO₃
- Description:4-Nitrophenol-2,3,5,6-D4