

4-Nitrophenol-[2,3,5,6-D₄] (98% D), Sigma-Aldrich®
Catalog # SIAL491608-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
ADR:6.1,III
:M-Clarity™ quality level = MQ200
UN:1663
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:93951-79-2
Formula:C₆HD₄NO₃
MDL Number:MFCD00143352