

4-Nitrophenol-[2,3,5,6-D₄] (98% D)
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
CAS Number:93951-79-2
:
Formula:C₆HD₄NO₃
MDL Number:MFCD00143352
Storage Temperature:Ambient
UN:1663
ADR:6.1,III
:GHS08
:GHS07
Frequently Bought Together