Order Entry
Czech Republic
ContactUsLinkComponent
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 99%
Irritant
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 99%
Certificates
undefined
Irritant
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 99%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    145 to 160 °C (Dec)
  • HPLC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together