(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:156311-83-0
:
Formula:C₁₇H₂₇F₆N₇OP₂
MDL Number:MFCD03703417
:521.4 g/mol
:M-Clarity™ quality level = MQ100
Frequently Bought Together