ContactUsLinkComponent
Shakers, rocker 2D and 3D basic and digital
 :  IKA
undefined
Shakers, rocker 2D and 3D basic and digital
 :  IKA
  • Catalog No:
  • 444-0817
  • 444-0816
  • 444-0815
  • 444-0818
  • Model:
  • Rocker 3D basic
  • Rocker 2D digital
  • Rocker 2D basic
  • Rocker 3D digital
  • Shaking motion:
  • Tumbling
  • Rocking
  • Rocking
  • Tumbling
  • Speed range:
  • 30 min⁻¹ (fixed)
  • 5 - 80 min⁻¹
  • 5 - 80 min⁻¹
  • 5 - 80 min⁻¹
  • Angle:
  • 0 - 15°
  • 0 - 15°
  • W×D×H:
  • 280×330×165 mm
  • 280×330×150 mm
  • 280×330×150 mm
  • 280×330×185 mm
  • Weight:
  • 2,2 kg
  • 2,2 kg
  • 2,2 kg
  • 2,2 kg
  • Max. load:
  • 2 kg
  • 2 kg
  • 2 kg
  • 2 kg
Frequently Bought Together