Cation exchange HPLC columns, ProPac™ SCX-10 and WCX-10
Supplier: Thermo Fisher Scientific
Certificates
About this item
The ProPac HPLC columns are weak and strong cation exchange columns with exceptionally high resolution and efficiency for separations of protein variants.
- Characterisation and quality control assessment of monoclonal antibodies and other proteins
- Useful for characterisation of related protein variants including deamidation and monoclonal antibodies lysine truncation variants
Based on stationary phases composed of 10 µm nonporous polymeric beads, these columns resolve isoforms that differ by a single charged residue. A thin, hydrophilic layer grafted to the particle core eliminates unwanted secondary interactions. The cation-exchange surface provides pH-based selectivity control and fast mass transfer for high-efficiency separation and moderate capacity.
Specifications
- Catalog No:
- 554-5302
- 555-0256
- 555-7454
- 555-7443
- 554-5301
- 554-5304
- 554-5303
- 555-7301
- 555-7289
- 555-7274
- 554-5300
- 554-5305
- 555-7453
- 555-7442
- 555-7441
- 555-7290
- Max. pressure:
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- pH range:
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- Temperature limit:
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
Specifications
Frequently Bought Together