Ion exchange SPE, StratoSpheres™
: VARIAN
This macroporous polystyrene product has a much higher loading compared to conventional functionalised silica SPE media and is compatible with all polar and non-polar, protic and aprotic solvents.
The freebase generation of an isolated organic TFA salt can be easily facilitated using PL-HCO₃ MP SPE tubes, used to remove TFA from HPLC solvent systems. Fractions from a preparative HPLC system can be passed through a PL-HCO₃ MP SPE tube and will effectively neutralise 0,1% TFA solutions from water, acetonitrile and combinations of both solvents.
Catch and release purification of TFA salts done by using PL-SO₃H MP SPE.
Frequently Bought Together