Sleeves for ground joints, Azlon®
Dodavatel: SciLabware
Certifikáty
O tomto produktu
Tapered sleeves, made of PTFE, for grease free and high vacuum seals using ground glass joints.
- Chemical resistance
- Forms a liquid and gas tight seal
- Self-lubricating and non contaminating
- Strong construction with knurled shoulder
Working temperature: −100 to +260 °C.
Working vacuum: ≤10 to 3 atmospheres.
Certifications: Manufactured from USP Class VI grade PTFE, suitable for food contact applications in accordance with USA regulations.
Frequently Bought Together