Order Entry
Czech Republic
ContactUsLinkComponent
Sanitary cleaners, Tana Professional
Sanitary cleaners, Tana Professional
Catalog # TANA713469
Supplier:  WERNER MERTZ
Sanitary cleaners, Tana Professional
Catalog # TANA713469
Supplier:  WERNER MERTZ

Specifications

  • Description:
    SANET spray
  • Packed:
    750 ml spray bottle
  • Pk:
    750 mL
  • pH value:
    2

Specifications

About this item

A range of cleaners suitable for most maintenance cleaning tasks in rest rooms.

green care® products are designed in accordance with the principles of sustainable development.

Certifications: European Ecolabel.