Capillary columns, TraceGOLD™ TG-1301MS
Dodavatel: Thermo Fisher Scientific
Certifikáty
O tomto produktu
TraceGOLD™ TG-1301MS columns have excellent reproducibility and column-to-column consistency even with sensitive detectors like ECDs and MS.
- Low bleed
- Low to mid-polarity phase, 6% cyanopropylphenyl/94% dimethyl polysilo
- Long lifetime
- Excellent inertness
- Equivalent to USP G43 phase
Frequently Bought Together