Order Entry
Czech Republic
ContactUsLinkComponent
Bottles, wide neck, square, HDPE, with screw caps
Bottles, wide neck, square, HDPE, with screw caps
  SEMA3047
 :  SEMADENI
undefined
Bottles, wide neck, square, HDPE, with screw caps
  SEMA3047
 :  SEMADENI

 

  • Ext.Ø:
    60 mm
  • Capacity:
    250 ml
  • Height:
    95 mm
  • Cap colour:
    Black
  • Neck I-Ø:
    43 mm
  • Pk:
    100

 

 

HDPE, translucent, with black screw cap.

  • Good chemical resistance
  • Good temperture resistance