Order Entry
Czech Republic
ContactUsLinkComponent
Buckets, Round, PP
Buckets, Round, PP
Katalogové číslo 216-1258
Dodavatel:  SEMADENI
undefined
Buckets, Round, PP
Katalogové číslo 216-1258
Dodavatel:  SEMADENI

Specifikace

  • Capacity:
    15 L
  • Colour:
    Natural
  • Description:
    With graduation and handle
  • Height:
    335 mm
  • Pk:
    1
  • Ø ext.:
    310 mm

Specifikace

O tomto produktu

Universal multipurpose bucket, made of PP, with spout, reinforced rim and handle.

  • Graduated, autoclavable
  • Very good thermal resistance
  • Very good chemical resistance