Order Entry
Czech Republic
ContactUsLinkComponent
Recombinant desoxyribonuclease I
Recombinant desoxyribonuclease I
Supplier:  Merck
Certificates
Recombinant desoxyribonuclease I
Supplier:  Merck

Specifications

  • CAS No.:
    9003-98-9
  • Enzyme type:
    Recombinant
  • Storage conditions:
    Freezer
  • Enzyme name:
    Desoxyribonuclease I
  • Enzyme synonyms:
    DNase I

Specifications

Frequently Bought Together