4-Chlorophenylurea
Catalog # PROOMM0162.07
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:170.6 g/mol
EINECS:205-412-6
CAS Number:140-38-5
Formula:C₇H₇ClN₂O
MDL Number:MFCD00014788