Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, 5-tert-Butyl-2,4,6-trinitro-m-xylene 10 µg/ml in cyclohexane
Catalog # PROONE0518
CAS Number:  
Organic reference standard, 5-tert-Butyl-2,4,6-trinitro-m-xylene 10 µg/ml in cyclohexane
Catalog # PROONE0518
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4,5 mL
  • CAS No.:
    81-15-2
  • Description:
    5-tert-Butyl-2,4,6-trinitro-m-xylene 10 µg/ml in cyclohexane
  • Formula:
    C₁₂H₁₅N₃O₆
  • Description:
    5-tert-Butyl-2,4,6-trinitro-m-xylene

Specifications

About this item