Sodium acetate ∼3 mol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIAL71196-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:82.03 g/mol
Storage Temperature:Ambient
EINECS:204-823-8
CAS Number:127-09-3
Formula:H₃CCOONa
MDL Number:MFCD00012459